The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012115.1 | NP_001012115.1 | 6 - 7 | 892 | 83 |
GenBank mRNA | BC089877.1 | - | 6 - 7 | 892 | 83 |