Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rexo1
Entrez Gene ID
314630
Gene Name
REX1, RNA exonuclease 1 homolog (S. cerevisiae)
Gene Aliases
Tceb3bp1
Chromosome Location
Chr.7: 12022268 - 12041660 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012114.2 NP_001012114.2 14 - 15 3524 72
GenBank mRNA BC083596.1 - 10 - 11 1519 72

View Full Product Details