Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f5
Entrez Gene ID
286905
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 5
Gene Aliases
-
Chromosome Location
Chr.7: 14559806 - 14574345 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173124.1 NP_775147.1 - 597 190
XM_006241051.2 XP_006241113.1 5 - 6 666 190
XM_008765158.1 XP_008763380.1 5 - 6 668 190
GenBank mRNA BC129085.1 - 5 - 6 558 190
U39207.1 - - 597 190

Gene Symbol
LOC100912471
Entrez Gene ID
100912471
Gene Name
cytochrome P450 4F5-like
Gene Aliases
-
Chromosome Location
Chr.7: 14760053 - 14830055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241097.2 XP_006241159.1 3 - 4 233 190

Gene Symbol
LOC691320
Entrez Gene ID
691320
Gene Name
similar to cytochrome P450 4F1
Gene Aliases
-
Chromosome Location
Chr.7: 13881303 - 13889392 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241045.1 XP_006241107.1 2 - 3 200 190

View Full Product Details