Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wiz
Entrez Gene ID
314598
Gene Name
widely-interspaced zinc finger motifs
Gene Aliases
-
Chromosome Location
Chr.7: 14374358 - 14402638 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108064.1 NP_001101534.1 3 - 4 629 84
XM_008765168.1 XP_008763390.1 3 - 4 1092 84

View Full Product Details