Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2l1
Entrez Gene ID
304741
Gene Name
transcription factor CP2-like 1
Gene Aliases
Tcfcp2l1
Chromosome Location
Chr.13: 34610689 - 34671617 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107170.1 NP_001100640.1 13 - 14 1430 83

View Full Product Details