Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alas2
Entrez Gene ID
25748
Gene Name
5'-aminolevulinate synthase 2
Gene Aliases
-
Chromosome Location
Chr.X: 23167576 - 23187356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013197.1 NP_037329.1 6 - 7 1021 71
GenBank mRNA BC081857.1 - - 1042 71
D86297.1 - 6 - 7 1021 71

View Full Product Details