Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkd2
Entrez Gene ID
308068
Gene Name
naked cuticle homolog 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.1: 32054412 - 32085416 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107454.1 NP_001100924.1 4 - 5 401 68
XM_008758692.1 XP_008756914.1 7 - 8 2135 68
XM_008758693.1 XP_008756915.1 4 - 5 916 68

View Full Product Details