The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008363.1 | NP_001008364.1 | 5 - 6 | 688 | 150 |
GenBank mRNA | BC085928.1 | - | 5 - 6 | 688 | 150 |