The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013867.1 | NP_001013889.1 | 19 - 20 | 2616 | 74 |
XM_006248858.2 | XP_006248920.1 | 20 - 21 | 3443 | 74 | |
XM_006248859.2 | XP_006248921.1 | 20 - 21 | 3311 | 74 | |
XM_006248860.2 | XP_006248922.1 | 20 - 21 | 3431 | 74 | |
XM_006248861.2 | XP_006248923.1 | 19 - 20 | 3352 | 74 | |
GenBank mRNA | BC081935.1 | - | 19 - 20 | 2616 | 74 |