Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wipi2
Entrez Gene ID
288498
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
-
Chromosome Location
Chr.12: 13969704 - 13998134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007615.1 NP_001007616.1 11 - 12 1424 85
GenBank mRNA BC079184.1 - 11 - 12 1424 85

View Full Product Details