Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prkar1b
- Entrez Gene ID
-
25521
- Gene Name
- protein kinase, cAMP dependent regulatory, type I, beta
- Gene Aliases
-
RGD1563094, RIbeta
- Chromosome Location
-
Chr.12: 17614472 - 17713567 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ212815.1 |
- |
- |
279 |
65 |
View Full Product Details