Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arpc1a
Entrez Gene ID
81824
Gene Name
actin related protein 2/3 complex, subunit 1A
Gene Aliases
-
Chromosome Location
Chr.12: 11272609 - 11294084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031146.2 NP_112408.1 2 - 3 216 91
GenBank mRNA AF315378.1 - 1 - 2 101 91
BC059131.1 - 2 - 3 196 91
DY310102.1 - 2 - 3 216 91
FQ214345.1 - 2 - 3 190 91

View Full Product Details