Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igf2bp3
Entrez Gene ID
312320
Gene Name
insulin-like growth factor 2 mRNA binding protein 3
Gene Aliases
Ab2-255, RGD1306512
Chromosome Location
Chr.4: 78744689 - 78879459 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749741.3 XP_003749789.1 3 - 4 569 94
XM_006236474.2 XP_006236536.1 3 - 4 443 94
XM_008762933.1 XP_008761155.1 3 - 4 204 94
GenBank mRNA AY724479.1 - 3 - 4 533 94

View Full Product Details