Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crhr2
Entrez Gene ID
64680
Gene Name
corticotropin releasing hormone receptor 2
Gene Aliases
Crf2r
Chromosome Location
Chr.4: 85286371 - 85329374 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022714.1 NP_073205.1 11 - 12 1310 152
XM_006236556.2 XP_006236618.1 12 - 13 1300 152
XM_006236557.2 XP_006236619.1 12 - 13 1297 152
XM_006236558.1 XP_006236620.1 11 - 12 1328 152
GenBank mRNA U16253.1 - 11 - 12 1310 152

View Full Product Details