The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109236.1 | NP_001102706.1 | 3 - 4 | 605 | 100 |
XM_008762910.1 | XP_008761132.1 | 3 - 4 | 879 | 100 |