Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm5
Entrez Gene ID
306222
Gene Name
LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.4: 87016003 - 87019219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107289.1 NP_001100759.1 2 - 3 195 152

View Full Product Details