Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt11
Entrez Gene ID
311952
Gene Name
:polypeptide N-acetylgalactosaminyltransferase 11
Gene Aliases
-
Chromosome Location
Chr.4: 6297255 - 6330765 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271319.1 NP_001258248.1 6 - 7 1008 71
NM_199393.2 NP_955425.2 5 - 6 925 71
XM_006235922.2 XP_006235984.1 4 - 5 1066 71
XM_006235923.2 XP_006235985.1 5 - 6 1418 71
GenBank mRNA BC062004.1 - 5 - 6 859 71
FQ223626.1 - 6 - 7 1008 71

View Full Product Details