Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mnx1
Entrez Gene ID
682076
Gene Name
motor neuron and pancreas homeobox 1
Gene Aliases
Hlxb9
Chromosome Location
Chr.4: 2376350 - 2381308 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271274.1 NP_001258203.1 2 - 3 892 63

View Full Product Details