Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glra4
Entrez Gene ID
367912
Gene Name
glycine receptor, alpha 4
Gene Aliases
RGD1564103
Chromosome Location
Chr.X: 107422357 - 107442878 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191914.1 NP_001178843.1 5 - 6 588 101

View Full Product Details