Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tceal6
Entrez Gene ID
679974
Gene Name
transcription elongation factor A (SII)-like 6
Gene Aliases
-
Chromosome Location
Chr.2: 102369895 - 102371893 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749245.3 XP_003749293.1 2 - 3 243 131
GenBank mRNA FQ213604.1 - 2 - 3 169 131

Gene Symbol
Tceal3
Entrez Gene ID
501628
Gene Name
transcription elongation factor A (SII)-like 3
Gene Aliases
RGD1562504
Chromosome Location
Chr.X: 107322085 - 107324054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_577026.6 XP_577026.4 2 - 3 231 131
GenBank mRNA FQ213674.1 - 2 - 3 208 131
FQ213749.1 - 2 - 3 169 131

View Full Product Details