Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc6
Entrez Gene ID
361771
Gene Name
zinc finger, DHHC-type containing 6
Gene Aliases
-
Chromosome Location
Chr.1: 276291411 - 276309633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037652.1 NP_001032741.1 - 190 84
GenBank mRNA AY886524.1 - - 71 84
BC107928.1 - - 190 84

View Full Product Details