Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam45a
Entrez Gene ID
308009
Gene Name
family with sequence similarity 45, member A
Gene Aliases
RGD1308326
Chromosome Location
Chr.1: 282188074 - 282211588 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127681.1 NP_001121153.1 5 - 6 659 92
XM_006231682.1 XP_006231744.1 5 - 6 651 92
GenBank mRNA BC167081.1 - 5 - 6 628 92

View Full Product Details