Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpn1
Entrez Gene ID
365466
Gene Name
carboxypeptidase N, polypeptide 1
Gene Aliases
Cpn
Chromosome Location
Chr.1: 263733887 - 263762758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053526.2 NP_445978.1 4 - 5 957 59
GenBank mRNA AB042599.1 - 4 - 5 966 59
BC088124.1 - 4 - 5 957 59
FQ219519.1 - 2 - 3 242 59

View Full Product Details