The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139193.3 | NP_631932.3 | 1 - 1 | 1343 | 123 |
GenBank mRNA | S77867.1 | - | 1 - 1 | 1343 | 123 |