The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013153.1 | NP_001013171.1 | 2 - 3 | 799 | 97 |
XM_008760420.1 | XP_008758642.1 | 2 - 3 | 286 | 97 | |
XM_008760421.1 | XP_008758643.1 | 2 - 3 | 907 | 97 | |
GenBank mRNA | BC079260.1 | - | 2 - 3 | 799 | 97 |