Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dntt
Entrez Gene ID
294051
Gene Name
DNA nucleotidylexotransferase
Gene Aliases
-
Chromosome Location
Chr.1: 260289626 - 260321174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012461.1 NP_001012479.1 6 - 7 889 131
GenBank mRNA BC089904.1 - 6 - 7 889 131

View Full Product Details