Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Flvcr1
Entrez Gene ID
683955
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases
-
Chromosome Location
Chr.13: 109578011 - 109629588 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769906.1 XP_008768128.1 3 - 4 1161 127

View Full Product Details