Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pvrl4
Entrez Gene ID
498281
Gene Name
poliovirus receptor-related 4
Gene Aliases
RGD1559826
Chromosome Location
Chr.13: 89755665 - 89774185 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109076.1 NP_001102546.1 8 - 9 1774 81

View Full Product Details