Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvrl4
Entrez Gene ID
498281
Gene Name
poliovirus receptor-related 4
Gene Aliases
RGD1559826
Chromosome Location
Chr.13: 89755665 - 89774185 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006250256.2 XP_006250318.1 7 - 8 1666 80
GenBank mRNA FQ228572.1 - 6 - 7 1217 80
FQ228642.1 - 6 - 7 1217 80

View Full Product Details