The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105969.1 | NP_001099439.1 | 8 - 9 | 1896 | 58 |
XM_006250276.2 | XP_006250338.1 | 9 - 10 | 2022 | 58 | |
XM_006250277.2 | XP_006250339.1 | 7 - 8 | 1811 | 58 |