Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcer1g
Entrez Gene ID
25441
Gene Name
Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide
Gene Aliases
-
Chromosome Location
Chr.13: 89601894 - 89606326 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131001.1 NP_001124473.1 4 - 5 285 235
GenBank mRNA BC160864.1 - 4 - 5 285 235

View Full Product Details