Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ufc1
Entrez Gene ID
445268
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
ufc1-s
Chromosome Location
Chr.13: 89661763 - 89668513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006250251.2 XP_006250313.1 1 - 2 89 94

View Full Product Details