Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Traf5
Entrez Gene ID
684173
Gene Name
TNF receptor-associated factor 5
Gene Aliases
-
Chromosome Location
Chr.13: 110758556 - 110801869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769908.1 XP_008768130.1 4 - 5 765 64

View Full Product Details