Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slamf6
Entrez Gene ID
498287
Gene Name
SLAM family member 6
Gene Aliases
RGD1561848
Chromosome Location
Chr.13: 90300619 - 90321781 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191932.1 NP_001178861.1 3 - 4 687 92
XM_006250301.2 XP_006250363.1 3 - 4 1108 92

View Full Product Details