Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf6
Entrez Gene ID
498287
Gene Name
SLAM family member 6
Gene Aliases
RGD1561848
Chromosome Location
Chr.13: 90300619 - 90321781 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191932.1 NP_001178861.1 6 - 7 920 174

View Full Product Details