Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tatdn3
Entrez Gene ID
289378
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.13: 109655292 - 109669573 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105987.1 NP_001099457.1 - 203 79

View Full Product Details