Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Opn3
Entrez Gene ID
498289
Gene Name
opsin 3
Gene Aliases
-
Chromosome Location
Chr.13: 93716481 - 93746994 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191933.1 NP_001178862.1 2 - 3 776 117

View Full Product Details