Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh26
Entrez Gene ID
311693
Gene Name
cadherin 26
Gene Aliases
-
Chromosome Location
Chr.3: 173969019 - 174020867 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191745.1 NP_001178674.1 8 - 9 1213 85
XM_008762509.1 XP_008760731.1 8 - 9 1467 85

View Full Product Details