Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phactr3
Entrez Gene ID
362284
Gene Name
phosphatase and actin regulator 3
Gene Aliases
Scapin1, scapinin
Chromosome Location
Chr.3: 173798542 - 173871386 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214459.2 NP_999624.1 3 - 4 432 81
GenBank mRNA AY500157.1 - 3 - 4 432 81

View Full Product Details