Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmeb2
Entrez Gene ID
83635
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 176756006 - 176791960 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031803.1 NP_113991.1 4 - 5 461 62
GenBank mRNA AF059273.1 - 4 - 5 461 62
AF205778.1 - - 355 62
AF205779.1 - - 355 62
AF205780.1 - - 355 62

View Full Product Details