Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx8
Entrez Gene ID
288504
Gene Name
sorting nexin 8
Gene Aliases
-
Chromosome Location
Chr.12: 16340922 - 16389500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105912.1 NP_001099382.1 2 - 3 312 102
XM_006248920.2 XP_006248982.1 2 - 3 307 102
XM_006248921.2 XP_006248983.1 2 - 3 707 102
XM_006248923.2 XP_006248985.1 2 - 3 580 102

View Full Product Details