Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdelr2
Entrez Gene ID
304290
Gene Name
KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2
Gene Aliases
-
Chromosome Location
Chr.12: 13192452 - 13210758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013122.1 NP_001013140.1 3 - 4 447 78
GenBank mRNA BC085786.1 - 3 - 4 447 78

View Full Product Details