Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stap1
Entrez Gene ID
305269
Gene Name
signal transducing adaptor family member 1
Gene Aliases
RGD1311584, Stap-1
Chromosome Location
Chr.14: 23576084 - 23604983 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025115.1 NP_001020286.1 7 - 8 759 146
GenBank mRNA BC091249.1 - 7 - 8 759 146

View Full Product Details