Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss11c
Entrez Gene ID
408213
Gene Name
transmembrane protease, serine 11c
Gene Aliases
Hatl3
Chromosome Location
Chr.14: 23405717 - 23461384 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003979.1 NP_001003979.1 1 - 2 50 145

View Full Product Details