Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lphn3
Entrez Gene ID
170641
Gene Name
latrophilin 3
Gene Aliases
Cirl3, Lec3
Chromosome Location
Chr.14: 28362176 - 28856419 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130822.1 NP_570835.1 10 - 11 2176 61
XM_008769968.1 XP_008768190.1 10 - 11 2280 61
XM_008769969.1 XP_008768191.1 10 - 11 2265 61
XM_008769970.1 XP_008768192.1 10 - 11 2280 61
XM_008769971.1 XP_008768193.1 10 - 11 2280 61
XM_008769973.1 XP_008768195.1 10 - 11 2280 61
XM_008769974.1 XP_008768196.1 10 - 11 2280 61
XM_008769975.1 XP_008768197.1 10 - 11 2280 61
XM_008769976.1 XP_008768198.1 10 - 11 2280 61
XM_008769977.1 XP_008768199.1 9 - 10 2076 61
XM_008769979.1 XP_008768201.1 9 - 10 2076 61
XM_008769980.1 XP_008768202.1 10 - 11 2280 61
XM_008769981.1 XP_008768203.1 10 - 11 2280 61
XM_008769982.1 XP_008768204.1 10 - 11 2279 61
XM_008769983.1 XP_008768205.1 10 - 11 2279 61
XM_008769984.1 XP_008768206.1 10 - 11 2279 61
XM_008769985.1 XP_008768207.1 9 - 10 2075 61
XM_008769986.1 XP_008768208.1 9 - 10 2075 61
GenBank mRNA AF063103.1 - 10 - 11 2176 61
AF081154.1 - 9 - 10 1997 61
AF081155.1 - 9 - 10 1997 61
AF081156.1 - 9 - 10 1997 61
AF081157.1 - 10 - 11 2201 61
AF081158.1 - 10 - 11 2201 61
AF081159.1 - 10 - 11 2201 61

View Full Product Details