Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ugdh
Entrez Gene ID
83472
Gene Name
UDP-glucose 6-dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.14: 44479614 - 44502845 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031325.1 NP_112615.1 2 - 3 269 129
GenBank mRNA AB013732.1 - 2 - 3 269 129

View Full Product Details