The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113776.1 | NP_001107248.1 | 2 - 3 | 389 | 76 |
XM_006250998.2 | XP_006251060.1 | 2 - 3 | 484 | 76 | |
GenBank mRNA | BC158872.1 | - | 2 - 3 | 389 | 76 |