Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lias
Entrez Gene ID
305348
Gene Name
lipoic acid synthetase
Gene Aliases
-
Chromosome Location
Chr.14: 44507217 - 44524289 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012037.1 NP_001012037.1 6 - 7 792 100
GenBank mRNA BC083708.1 - 6 - 7 792 100

View Full Product Details