Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lnx1
Entrez Gene ID
360926
Gene Name
ligand of numb-protein X 1, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.14: 36047120 - 36149908 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108358.1 NP_001101828.1 6 - 7 1555 116
XM_006250902.2 XP_006250964.1 6 - 7 1549 116
XM_006250903.2 XP_006250965.1 5 - 6 3587 116
XM_006250904.2 XP_006250966.1 5 - 6 3557 116
GenBank mRNA EU219404.1 - 5 - 6 1398 116
EU219405.1 - 5 - 6 1398 116
EU219406.1 - 5 - 6 1266 116

View Full Product Details