Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss11b
Entrez Gene ID
365265
Gene Name
transmembrane protease, serine 11b
Gene Aliases
Hatl5, Tmprss11bnl
Chromosome Location
Chr.14: 24103869 - 24125260 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770049.1 XP_008768271.1 1 - 2 108 64

View Full Product Details